Learn More
COLDER PRODUCTS COMPANY MALE COUPLING 3/8 250PK
Supplier:  COLDER PRODUCTS COMPANY MPC22006T39M
Encompass_Preferred
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Insert Polysulfone w/Platinum Cured Silicone Seal Diameter Inner 9 5 mm 0 37 in Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber
Catalog No. 50-313-1448