Learn More
COLDER PRODUCTS COMPANY MALE INSERT 3/8 ID HB 25/PK
Supplier: COLDER PRODUCTS COMPANY PLCD2200612
CPC Plastic Connectors Couplings and Fittings Polypropylene Description 1 35 L x 0 63 in H (34 2 x 16 5 mm) Hose Barb Valved In-line Coupling Insert White For Use With 9 5mm Includes Plastic connector Overall Length 1 35 in 34 2 mm Height 0 63 in 16 5 mm Maximum Pressure 120 psi