Learn More
COLDER PRODUCTS COMPANY MALE INSERT 3/8 ID HB 25/PK
Catalog No. 503131279
Encompass_Preferred
CPC Plastic Connectors Couplings and Fittings Buna-N Seal Description 1 91 L x 0 94 in H (48 x 23 mm) Hose Barb Non-Valved Panel Mount Coupling Insert 0 25 in insert x 0 375 in hose barb White For Use With 9 5mm Includes Plastic connector Overall Length 1 91 in 48 mm
Catalog No. 50-313-1279 Supplier COLDER PRODUCTS COMPANY Supplier No. PLC42006
CPC Plastic Connectors Couplings and Fittings Buna-N Seal Description 1 91 L x 0 94 in H (48 x 23 mm) Hose Barb Non-Valved Panel Mount Coupling Insert 0 25 in insert x 0 375 in hose barb White For Use With 9 5mm Includes Plastic connector Overall Length 1 91 in 48 mm