Learn More
COLDER PRODUCTS COMPANY MALE INSERT 3/8 ID HB 25/PK
Catalog No. 503131355
Encompass_Preferred
CPC Plastic Connectors Couplings and Fittings Acetal Description 1 91 L x 0 94 in H (48 x 23 mm) Hose Barb Valved Panel Mount Coupling Insert White For Use With 9 5mm Includes Plastic connector Overall Length 1 91 in 48 mm Height 0 94 in 23 mm Maximum Pressure 120 psi
Catalog No. 50-313-1355 Supplier COLDER PRODUCTS COMPANY Supplier No. PLCD42006
CPC Plastic Connectors Couplings and Fittings Acetal Description 1 91 L x 0 94 in H (48 x 23 mm) Hose Barb Valved Panel Mount Coupling Insert White For Use With 9 5mm Includes Plastic connector Overall Length 1 91 in 48 mm Height 0 94 in 23 mm Maximum Pressure 120 psi