Learn More
COLDER PRODUCTS COMPANY MALE INSERT 5/16 ID HB 25/PK
Catalog No. 503131307
Encompass_Preferred
CPC Plastic Connectors Couplings and Fittings Acetal Description 1 35 L x 0 62 in (34 2 x 16 5 mm) Hose Barb Valved In-line Coupling Insert White For Use With 8mm Includes Plastic connector Overall Length 1 35 in 34 2 mm Height 0 62 in 16 5 mm Maximum Pressure 120 psi
Catalog No. 50-313-1307 Supplier COLDER PRODUCTS COMPANY Supplier No. PLCD22005
CPC Plastic Connectors Couplings and Fittings Acetal Description 1 35 L x 0 62 in (34 2 x 16 5 mm) Hose Barb Valved In-line Coupling Insert White For Use With 8mm Includes Plastic connector Overall Length 1 35 in 34 2 mm Height 0 62 in 16 5 mm Maximum Pressure 120 psi