Learn More
COLDER PRODUCTS COMPANY MALE INSRT 1/4 ID HB EL 25/PK
Catalog No. 503131306
Encompass_Preferred
CPC Plastic Connectors Couplings and Fittings Buna-N Seal Description 1 1 L x 0 67 in H (28 x 16 5 mm) Hose Barb Non-Valved Elbow Coupling Insert White For Use With 6 4mm Includes Plastic connector Overall Length 1 1 in 28 mm Height 0 67 in 16 5 mm Maximum Pressure 120 psi
Catalog No. 50-313-1306 Supplier COLDER PRODUCTS COMPANY Supplier No. PLC23004
CPC Plastic Connectors Couplings and Fittings Buna-N Seal Description 1 1 L x 0 67 in H (28 x 16 5 mm) Hose Barb Non-Valved Elbow Coupling Insert White For Use With 6 4mm Includes Plastic connector Overall Length 1 1 in 28 mm Height 0 67 in 16 5 mm Maximum Pressure 120 psi