Learn More
COLDER PRODUCTS COMPANY MALE INSRT 1/4 ID HB VAL 25PK
Catalog No. 503131436
Encompass_Preferred
CPC Plastic Connectors Couplings and Fittings Buna-N Seal Description 1 91 L x 0 94 in H (48 x 23 mm) Hose Barb Valved Panel Mount Coupling Insert White For Use With 6 4mm Includes Plastic connector Overall Length 1 91 in 48 mm Height 0 94 in 23 mm Maximum Pressure 120 psi
Catalog No. 50-313-1436 Supplier COLDER PRODUCTS COMPANY Supplier No. PLCD42004
CPC Plastic Connectors Couplings and Fittings Buna-N Seal Description 1 91 L x 0 94 in H (48 x 23 mm) Hose Barb Valved Panel Mount Coupling Insert White For Use With 6 4mm Includes Plastic connector Overall Length 1 91 in 48 mm Height 0 94 in 23 mm Maximum Pressure 120 psi