Learn More
COLDER PRODUCTS COMPANY MALE INSRT 3/8 ID HB EL 25/PK
Catalog No. 503131421
Encompass_Preferred
CPC Plastic Connectors Couplings and Fittings Acetal Description 1 25 L x 0 67 in H (31 7 x 16 5 mm) Hose Barb Valved Elbow Coupling Insert White For Use With 9 5mm Includes Plastic connector Overall Length 1 25 in 31 7 mm Height 0 67 in 16 5 mm Maximum Pressure 120 psi
Catalog No. 50-313-1421 Supplier COLDER PRODUCTS COMPANY Supplier No. PLCD23006
CPC Plastic Connectors Couplings and Fittings Acetal Description 1 25 L x 0 67 in H (31 7 x 16 5 mm) Hose Barb Valved Elbow Coupling Insert White For Use With 9 5mm Includes Plastic connector Overall Length 1 25 in 31 7 mm Height 0 67 in 16 5 mm Maximum Pressure 120 psi