Learn More
Nordson Medical MALE LR LOCK RING 1/16 1000PK
Catalog No. NC2120863
Encompass_Preferred
Male Luer Integral Lock Ring to 200 Series Barb, 1/16" (1.6 mm) ID Tubing, natural kynar pvdf 1000pk
This item is not returnable. View return policy
Catalog No. NC2120863 Supplier Nordson Medical Supplier No. MTLL210J2A
Male Luer Integral Lock Ring to 200 Series Barb, 1/16" (1.6 mm) ID Tubing, natural kynar pvdf 1000pk