Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polycarbonate, Straight, Male Large Bore Luer to Hose Barb Adapter, 1/4"" ID; 25/PK
Catalog No. NC1280043
Encompass
These large-bore adapters are perfect for handling higher flow volumes in process while still using a slip-lock-style fitting system (similar to luer-style fittings). Adapters are manufactured in an ISO Class 8 (formerly Class 100,000) environment, are inspected to be free of contaminants, and are fully lot traceable. Packs of 25. Polycarbonate: Maximum temperature: 266¿F (130¿C). Sterilization: Gamma irradiation, ETO. Maximum temperature: 252¿F (122¿C). Sterilization: Gamma irradiation, ETO. Nylon: Maximum temperature: 194¿F (90¿C). Sterilization: Gamma irradiation, ETO, autoclave.
Catalog No. NC1280043 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4550952
These large-bore adapters are perfect for handling higher flow volumes in process while still using a slip-lock-style fitting system (similar to luer-style fittings). Adapters are manufactured in an ISO Class 8 (formerly Class 100,000) environment, are inspected to be free of contaminants, and are fully lot traceable. Packs of 25. Polycarbonate: Maximum temperature: 266¿F (130¿C). Sterilization: Gamma irradiation, ETO. Maximum temperature: 252¿F (122¿C). Sterilization: Gamma irradiation, ETO. Nylon: Maximum temperature: 194¿F (90¿C). Sterilization: Gamma irradiation, ETO, autoclave.