Learn More
AVANTOR MASTERFLEX (VWR LLC) Cadence Luer Fitting Adapter, nickel-plated brass, male luer lock to 1/8"" NPT(M); 1/ea
Catalog No. NC0595361
Encompass
Exceptional durability. Easy to disassemble for cleaning. Autoclavable.
Catalog No. NC0595361 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4150784
Exceptional durability. Easy to disassemble for cleaning. Autoclavable.