Learn More
S4J MANUFACTURING SERVICES INC MALE LUER LOCK (9/16" HEX)
Catalog No. NC2110976
Encompass
Male Luer Lock (9/16" hex), 1/4" NPT male. Tapered Pipe Thread: 1/4" NPT male thread. Material: Plated Brass
This item is not returnable. View return policy
Catalog No. NC2110976 Supplier S4J MANUFACTURING SERVICES INC Supplier No. A1304
Male Luer Lock (9/16" hex), 1/4" NPT male. Tapered Pipe Thread: 1/4" NPT male thread. Material: Plated Brass