Learn More
SAI Infusion Technologies Piercable Injection Cap with Male Luer
SDP
Catalog No. NC9080409
Encompass
Male luer lock injection site - bi-directional , latex free polycarbonate, polyisoprene. Sterile, 10/box. Note: access with a standard needle may cause coring, Huber needle is recommended (such as a H22-75, sold separately). Product is for laboratory animal research only.
Catalog No. NC9080409 Supplier SAI Infusion Technologies Supplier No. IC
Male luer lock injection site - bi-directional , latex free polycarbonate, polyisoprene. Sterile, 10/box. Note: access with a standard needle may cause coring, Huber needle is recommended (such as a H22-75, sold separately). Product is for laboratory animal research only.