Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Straight, Male Luer Lock x Plug; 25/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4550556
Encompass
Easily complete your luer fittings systems with these premium connectors. All female ports may be used with either slip or luer lock connections. Plugs and caps are also available.
Catalog No. NC9731091