Learn More
Chemglass Life Sciences Male Luer Lock to 1/4-5/16 in Hose Barb Adapter, 316 Stainless Steel
SDP
Catalog No. NC3673080
Encompass_Preferred
This stainless steel adapter connects a standard male luer lock fitting to flexible tubing via a 1/4-5/16 in hose barb, providing a secure, corrosion-resistant connection for laboratory fluid transfer and sampling applications.
  • Constructed from 316 stainless steel for corrosion resistance.
  • Connects male luer lock fittings to tubing with a 1/4-5/16 in hose barb.
  • Threaded/tabbed body provides a secure, leak-resistant connection.
  • Suitable for laboratory fluid transfer, sampling, and process applications.
  • Sold individually (each).
Catalog No. NC3673080 Supplier Chemglass Life Sciences Supplier No. CLS9010M250
This stainless steel adapter connects a standard male luer lock fitting to flexible tubing via a 1/4-5/16 in hose barb, providing a secure, corrosion-resistant connection for laboratory fluid transfer and sampling applications.
  • Constructed from 316 stainless steel for corrosion resistance.
  • Connects male luer lock fittings to tubing with a 1/4-5/16 in hose barb.
  • Threaded/tabbed body provides a secure, leak-resistant connection.
  • Suitable for laboratory fluid transfer, sampling, and process applications.
  • Sold individually (each).