Learn More
Cole-Parmer Luer to Plug Fitting, Straight Adapter, Natural Nylon, Cleanroom Packed, Male Luer Lock; 10/Pk
Supplier:  Cole-Parmer 5011052
Encompass_Preferred
These Cole-Parmer cleanroom fittings are high-quality PVC-free fittings, ideal in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. Simply connect the plug to a female luer adapter to stop the fluid flow. Plugs are available in five different formulations—select one that best suits your application.
Catalog No. 50-255-8175