Learn More
AVANTOR MASTERFLEX (VWR LLC) CrystalVu luer fitting, male luer lock x 1/4"" hose barb, Berry, 25/pk
Catalog No. NC1565973
Encompass
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet, and orange. Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA). Superior chemical resistance compared to polycarbonate. Sterilize by autoclave, gamma irradiation, or ethylene oxide. Meets USP Class VI. Animal-derived ingredient-free. Packs of 25.
Catalog No. NC1565973 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4552006
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet, and orange. Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA). Superior chemical resistance compared to polycarbonate. Sterilize by autoclave, gamma irradiation, or ethylene oxide. Meets USP Class VI. Animal-derived ingredient-free. Packs of 25.