Learn More
Braintree Scientific Male Luer Spring Tether,14"" w/catheter to fit 25g swivel
SDP
Catalog No. 501955242
Encompass
Male Spring Tether For researchers that wish to use a standard swivel or stainless steel swivel with the Quick Connect™ Harness simply use a tether extension line with intergraded male end. Individually packaged and ETO exposed. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5242 Supplier Braintree Scientific Supplier No. MST1425G
Male Spring Tether For researchers that wish to use a standard swivel or stainless steel swivel with the Quick Connect™ Harness simply use a tether extension line with intergraded male end. Individually packaged and ETO exposed. **Sold for research purposes only and not for human use**