Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Straight, Male Luer Lock to Hose Barb Adapter, 5/32"" ID; 25/PK
Catalog No. NC1495381
Encompass
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
Catalog No. NC1495381 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4550506
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.