Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight , Male Luer to Hose Barb Adapter, 1/16"" ID; 25/PK
Catalog No. NC0465155
Encompass
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.
Catalog No. NC0465155 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4551822
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.