Learn More
SAI Infusion Technologies Male to Male Luer Connector
SDP
Catalog No. NC9876577
Encompass
Polycarbonate Luer Connector combines two male luer fittings and allows connection of two extension lines with female luer ends. Connectors are ETO sterilized and come in a box of 10. Product is for laboratory animal research only.
Catalog No. NC9876577 Supplier SAI Infusion Technologies Supplier No. MML
Polycarbonate Luer Connector combines two male luer fittings and allows connection of two extension lines with female luer ends. Connectors are ETO sterilized and come in a box of 10. Product is for laboratory animal research only.