Learn More
Sigma Aldrich Male Mini Luer Plug Single Fluidic 334 Tpe
Catalog No. 9218741EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
92-187-41EA 1 Ea.
1 options
Catalog No. 92-187-41EA Supplier Sigma Aldrich Supplier No. 9218741EA
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
The male mini luer plugs are the means to close the female Mini Luer interfaces on our microfluidic platforms. The male mini luer plug is made of a soft thermoplastic elastomer (TPE). The soft material allows for easy closing of the interface without added pressure to the chip
Specifications
Quantity 1 Ea.