Learn More
Sigma Aldrich Male Mini Luer Plugs - Low Vol. Displ. Fluidic 438 Pp
Catalog No. 9222261EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
92-222-61EA 1 Ea.
1 options
Catalog No. 92-222-61EA Supplier Sigma Aldrich Supplier No. 9222261EA
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
These special male Mini Luer plugs are designed to fit female Mini Luer interfaces. They seal the connecting hole, while at the same time liquid movement is avoided due to minimal air or liquid displacement.
Specifications
Quantity 1 Ea.