Learn More
Gardner Denver Welch Vacuum Tech Inc MALE NPT PIPE INLET/EXHAUST
SDP
Catalog No. NC0228666
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
MALE NPT PIPE INLET/EXHAUSTFor Welch Pump. For Welch Pump Models 1380,1402,1376,1405 Male 3/4" NPT to 1"-20 Pump Pump Thread Size1" -20
Catalog No. NC0228666 Supplier Gardner Denver Welch Vacuum Tech Inc Supplier No. 1393P
This item has a minimum qty of 2 per supplier requirements.
MALE NPT PIPE INLET/EXHAUSTFor Welch Pump. For Welch Pump Models 1380,1402,1376,1405 Male 3/4" NPT to 1"-20 Pump Pump Thread Size1" -20