Learn More
Microsolv Technology Corporation Male Nut 2Pc Singl Ferule Peek
Catalog No. 5001451579
Encompass_Preferred
Tubing Connector Fittings, High Pressure, Two Pieces, 1/16th, PEEK, Natural, Large Head, Standard Length and 10-32 Screw Threads. Use with Single Cone Ferrules. Finger Tighten, MicroSolv Brand. 10/PK
Catalog No. 50-014-51579 Supplier Microsolv Technology Corporation Supplier No. 4732033
Tubing Connector Fittings, High Pressure, Two Pieces, 1/16th, PEEK, Natural, Large Head, Standard Length and 10-32 Screw Threads. Use with Single Cone Ferrules. Finger Tighten, MicroSolv Brand. 10/PK