Learn More
Microsolv Technology Corporation Male Nut Endure Fngtt Sm
Catalog No. 5001451514
Encompass_Preferred
Tubing Connector Fittings, High Pressure, One Piece, 1/16th, Carbon PEEK, Black, Small Head with 10-32 Screw Threads. Finger Tighten, Endure Brand. 10/PK.
Catalog No. 50-014-51514 Supplier Microsolv Technology Corporation Supplier No. 4730101
Tubing Connector Fittings, High Pressure, One Piece, 1/16th, Carbon PEEK, Black, Small Head with 10-32 Screw Threads. Finger Tighten, Endure Brand. 10/PK.