Learn More
Microsolv Technology Corporation Male Nut Long Body Peek
Catalog No. 5001451555
Encompass_Preferred
Tubing Connector Fittings, High Pressure, One Piece, 1/16th, PEEK, Natural, Standard Size Head but a Long Length with 10-32 Screw Threads. Finger Tighten, MicroSolv Brand. 1/EA.
Catalog No. 50-014-51555 Supplier Microsolv Technology Corporation Supplier No. 4732003
Tubing Connector Fittings, High Pressure, One Piece, 1/16th, PEEK, Natural, Standard Size Head but a Long Length with 10-32 Screw Threads. Finger Tighten, MicroSolv Brand. 1/EA.