Learn More
Microsolv Technology Corporation Male Nuts Combihead Flat Blue
Catalog No. 5001451565
Encompass_Preferred
Tubing Connector Fittings, High Pressure, One Piece, 1/16th, PEEK, Blue, Large Combihead with 10-32 Screw Threads. Finger or wrench Tighten, MicroSolv Brand. 10/PK.
Catalog No. 50-014-51565 Supplier Microsolv Technology Corporation Supplier No. 4732011B
Tubing Connector Fittings, High Pressure, One Piece, 1/16th, PEEK, Blue, Large Combihead with 10-32 Screw Threads. Finger or wrench Tighten, MicroSolv Brand. 10/PK.