Learn More
Microsolv Technology Corporation Male Nuts, Hplc, Peek 8Mm Head
Catalog No. NC1547193
Encompass_Preferred
Tubing Connector Fittings, High Pressure, One Piece, 1/16th, PEEK, Natural, Small, ""Combihead"" Hex Head with 10-32 Screw Threads. Finger Tighten, MicroSolv Brand. 10/PK.
Catalog No. NC1547193 Supplier Microsolv Technology Corporation Supplier No. 4732026
Tubing Connector Fittings, High Pressure, One Piece, 1/16th, PEEK, Natural, Small, ""Combihead"" Hex Head with 10-32 Screw Threads. Finger Tighten, MicroSolv Brand. 10/PK.