Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Compression to Threaded Adapter, 1/2"" OD x 1/2"" NPT(M); 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0638570
Encompass
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Catalog No. NC0916331