Learn More
Bellco Glass, Inc. Male Quick Turn Coupling For 1/4 ID Tubing
SDP
Catalog No. 501946382
Encompass_Preferred
Male Quick Turn Coupling ¼". Male plastic quick turn tube couplingMax. Pressure: 125psi @72°f. Material: clear polycarbonate. Temperature range: 200°f (continuous). Use with air & water. Material meets: ANSI/HIMA md 70.1 & iso 594/1-1986Tubing: use PVC, polyurethane, & silicone tubing with a durometer- of a75-a85Clamps: not requiredPolycarbonate: fair chemical, impact, strength & abrasion resistanceQTY/CASE: 10. MADE IN THE USA
Catalog No. 50-194-6382 Supplier Bellco Glass, Inc. Supplier No. 780800003
Male Quick Turn Coupling ¼". Male plastic quick turn tube couplingMax. Pressure: 125psi @72°f. Material: clear polycarbonate. Temperature range: 200°f (continuous). Use with air & water. Material meets: ANSI/HIMA md 70.1 & iso 594/1-1986Tubing: use PVC, polyurethane, & silicone tubing with a durometer- of a75-a85Clamps: not requiredPolycarbonate: fair chemical, impact, strength & abrasion resistanceQTY/CASE: 10. MADE IN THE USA