Learn More
Checkers Male Ramp for 5-Channel Yellow Jacket™ AMS Modular Accessibility System
Catalog No. 19088407 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Yellow
Catalog No. Color
19-088-407 Yellow
1 options
Catalog No. 19-088-407 Supplier Checkers Supplier No. WSA125RMAHDY
Only null left
Add to Cart
Add to Cart
The Yellow Jacket™ 5-channel AMS™ (Advanced Modular System) is a patented technology that utilizes separate cable protector sections and ramps to create a system that is fully expandable to hold an unlimited number of cables.
  • Protects cables and hoses up to 1.25 in. outside diameter
  • Connect unlimited center sections and add ramps
  • Hinged lid for easy cable placement
  • Patented technology with modular interlocking design
Specifications
Product Type Cable Protector Kit
Color Yellow
Material HDPE