Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Male Rotating Luer Lock and Nut Assembly with 1/16 in. ID - Polypropylene - QC
SDP

Catalog No. 01000248 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Berry Blue
Lime Green
Natural
Rose Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01-000-248 Lime Green
01-000-247 Berry Blue
01-000-251 Rose Red
01-000-253 Yellow
01-000-249 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-000-248
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Nut and body are locked and rotate as a single unit. Precision engineered for smooth positive sealing. Color nut for easy recognition. This standard luer is ISO 80369-7 compliant. 

  • Single & Multi Use Applications
  • PVC-Free
  • ISO 80369-7 Compliant
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661, USP81, USP 88
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty and Services

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Compliance Certificates Available
Description Class 7 Cleanroom Manufactured
Material Non-Animal Derived Polypropylene
For Use With (Application) Tubing connections; Medical device input/output ports; Drug and fluid transfer lines
Packaging Quantity 100 Pack
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Lime Green
Product Type Rotating Luer Lock
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.