Learn More
AVANTOR MASTERFLEX (VWR LLC) Fitting, PTFE, Straight, Threaded Plug, 1/4"" NPT(M)
Catalog No. NC1659163
Encompass
Very good chemical resistance. Rated to 200 psi at 77¿F ( 25¿C). Usable from -65 to 190¿F (-53 to 87¿C).
Catalog No. NC1659163 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3403056
Very good chemical resistance. Rated to 200 psi at 77¿F ( 25¿C). Usable from -65 to 190¿F (-53 to 87¿C).