Learn More
Grainger Male Thread Branch Tee 8mmx1/8 BSPT
Catalog No. 502764435
Encompass_Preferred
The Grainger Approved Male Thread Branch Tee is a 1/2" NPT male thread branch tee made of malleable iron. It has one 1/2" NPT male inlet and two 1/2" NPT female outlets in a tee configuration. This fitting allows you to branch off in two directions from the main line. It is rated for a maximum pressure of 300 PSI at 70°F and is commonly used in industrial compressed air, water, oil, natural gas, and steam applications. The Grainger Approved line offers quality fittings at an economical price point. This branch tee provides a durable and reliable option for creating branch connections in threaded piping systems.
Catalog No. 50-276-4435 Supplier Grainger Supplier No. 170K42
The Grainger Approved Male Thread Branch Tee is a 1/2" NPT male thread branch tee made of malleable iron. It has one 1/2" NPT male inlet and two 1/2" NPT female outlets in a tee configuration. This fitting allows you to branch off in two directions from the main line. It is rated for a maximum pressure of 300 PSI at 70°F and is commonly used in industrial compressed air, water, oil, natural gas, and steam applications. The Grainger Approved line offers quality fittings at an economical price point. This branch tee provides a durable and reliable option for creating branch connections in threaded piping systems.