Learn More
3B Scientific™ Male Urninary System Model, with 3B Smart Anatomy
SDPSCI_ED
Catalog No. VF325R Shop All 3B Scientific Products
Change view
Click to view available options
Size:
3/4 Life-size
Catalog No. Size
VF325R 3/4 Life-size
1 options
Catalog No. VF325R Supplier 3B Scientific™ Supplier No. VF325
Add to Cart
Add to Cart
3B Scientific™ Male Urninary System Model has anatomical detail that makes it ideal for classroom or doctor's offices. Not delivered on base.
Model shows the structures of the retroperitoneal cavity:
  • Right kidney is shown open
  • Inferior vena cava
  • Renal veins
  • Aorta with its branches
  • Iliacal vessels
  • Ureter
  • Urinary bladder
  • Prostate
  • Adrenal gland
  • Rectum
  • Musculature
Additional Features:
  • Anatomy app 3B Smart Anatomy included and includes 23 digital anatomy lectures, 117 different virtual anatomy models and 39 anatomy quizzes
  • Access to the anatomy course 3B Smart Anatomy, hosted inside the Complete Anatomy app by 3D4Medical
  • Warranty upgrade from 3 to 5 years with every product registration
  • Access to a free 3-day trial to all premium features of the Complete Anatomy app
Specifications
For Use With (Application) Studying anatomy
Includes Anatomy software
Size 3/4 Life-size