Learn More
Medchemexpress LLC Maleic acid | 110-16-7 | MFCD00063177 | >98.0% | 116.07 | C4H4O4 | 5g
SDP
Catalog No. 5000268228
Encompass_Preferred
Maleic Acid is a Glutamate Decarboxylase (GAD) inhibitor of E coli and L monocytogenes
Catalog No. 50-002-68228 Supplier Medchemexpress LLC Supplier No. HYY03675G
Maleic Acid is a Glutamate Decarboxylase (GAD) inhibitor of E coli and L monocytogenes