Learn More
Selleck Chemical LLC Maleic acid S3086-1g
SDP
Catalog No. 503110030
Encompass_Preferred
Maleic acid inhibits glutamate decarboxylase (GAD) activity thus enhances acid sensitivity of Listeria monocytogenes Maleic acid affects the extracellular GABA levels
Catalog No. 50-311-0030 Supplier Selleck Chemical LLC Supplier No. S30861G
Maleic acid inhibits glutamate decarboxylase (GAD) activity thus enhances acid sensitivity of Listeria monocytogenes Maleic acid affects the extracellular GABA levels