Learn More
Selleck Chemical LLC Maleic acid S3086-100mg
SDP
Catalog No. 503111569
Encompass_Preferred
Maleic acid inhibits glutamate decarboxylase (GAD) activity thus enhances acid sensitivity of Listeria monocytogenes Maleic acid affects the extracellular GABA levels
Catalog No. 50-311-1569 Supplier Selleck Chemical LLC Supplier No. S3086100MG
Maleic acid inhibits glutamate decarboxylase (GAD) activity thus enhances acid sensitivity of Listeria monocytogenes Maleic acid affects the extracellular GABA levels