Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maleic Acid, 99%

Catalog No. p-3734387
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125231000 100 g Plastic bottle
AC125230010 1 kg Plastic bottle
AC125230051 5 kg Plastic drum
AC125230250 25 kg Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC125231000 Supplier Thermo Scientific Chemicals Supplier No. 125231000
Add to Cart

CAS: 110-16-7 | C4H4O4 | 116.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-16-7
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00063177
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
Synonym maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
PubChem CID 444266
ChEBI CHEBI:18300
IUPAC Name (Z)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

Melting Point 134.0°C to 138.0°C
Color White
Assay Percent Range 99%
Linear Formula HO2CCH=CHCO2H
Beilstein 02, IV, 2199
Merck Index 15, 5767
Solubility Information Solubility in water: 790g/L (25°C). Other solubilities: soluble in glacial acetic acid, soluble in alcohol and acetone, slightly soluble in ether and benzene, practically insoluble in carbon tetrachloride
Formula Weight 116.07
Percent Purity 99%
Quantity 100 g
Flash Point 127°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Chemical Name or Material Maleic acid
Show More Show Less
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.