Learn More
Medchemexpress LLC Maleic acid-d2 | 24461-33-4 | MFCD00144720 | 99.3% | 118.08 | C4H2D2O4 | 5mg
SDP
Catalog No. 5000268219
Encompass_Preferred
Maleic Acid-d2 is the deuterium labeled Maleic Acid[1] Maleic Acid is a Glutamate Decarboxylase (GAD) inhibitor of E coli and L monocytogenes[2][3]
Catalog No. 50-002-68219 Supplier Medchemexpress LLC Supplier No. HYY0367S5MG
Maleic Acid-d2 is the deuterium labeled Maleic Acid[1] Maleic Acid is a Glutamate Decarboxylase (GAD) inhibitor of E coli and L monocytogenes[2][3]