Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maleic acid, disodium salt, 99%, Thermo Scientific™

Catalog No. AC215541000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC215541000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC215541000 Supplier Thermo Scientific Chemicals Supplier No. 215541000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-47-1
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
InChI Key VXXVUHAXJHEYFH-UAIGNFCESA-N
PubChem CID 87068830
IUPAC Name (Z)-but-2-enedioic acid;sodium
SMILES C(=CC(=O)O)C(=O)O.[Na].[Na]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 0.99
Linear Formula NaO2CCH=CHCO2Na
Quantity 100 g
Beilstein 02, 751
Formula Weight 160.04
Percent Purity ≥98.5%
Physical Form Amorphous Powder
Chemical Name or Material Maleic acid, disodium salt, 0.99
Hazard Category HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.