Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maleic Acid, NF, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18604146 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-146 25 g Amber Glass Bottle
18-604-147 100 g Amber Glass Bottle
18-604-149 500 g Amber Glass Bottle
18-604-148 12 kg Fiber Drum
18-604-145 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-146 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M120325GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

M1203, 110-16-7, C4H4O4

Spectrum™ Chemical Maleic Acid, NF

Chemical Identifiers

CAS 110-16-7
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00063177
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
IUPAC Name (2E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

CAS Max % 100%
UN Number UN3261
Percent Purity 99 to 101%
Grade NF
Quantity 25 g
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.