Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maleic anhydride, briquettes

Catalog No. AC465660251
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465660251 25 kg
AC465660250 25 g
AC465660010 1 kg
AC465660050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC465660251 Supplier Thermo Scientific Chemicals Supplier No. 465660251
Add to Cart

CAS: 108-31-6 | C4H2O3 | 98.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-31-6
Molecular Formula C4H2O3
Molecular Weight (g/mol) 98.06
MDL Number MFCD00005518
InChI Key FPYJFEHAWHCUMM-UHFFFAOYSA-N
IUPAC Name 2,5-dihydrofuran-2,5-dione
SMILES O=C1OC(=O)C=C1

Specifications

Melting Point 52.0°C to 55.0°C
Boiling Point 200.0°C
Flash Point 102°C
Quantity 25 kg
Solubility Information Solubility aq. soln.: 400 g/L (25°C). Other solubilities: reacts with water forming maleic acid, 227 g/100 g acetone - 50 g/100 g benzene (25°C), 112 g/100 g ethylacetate (25°C)- 0,60 g/100 g ccl4, 52.5 g/100 g chloroform (25°C)-23,4 g/100 g toluen
Viscosity 1.6 mPa.s (60°C)
Formula Weight 98.06
Chemical Name or Material Maleic anhydride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed.

Causes severe skin burns and eye damage.

May cause an allergic skin reaction.

May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Causes damage to organs through
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF SWALLOWED: Rinse mouth.

Do NOT induce vomiting.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower Immediately call a POISON CENTER or doctor/physician.

IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.

EINECSNumber 203-571-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.