Learn More
Medchemexpress LLC Maleic hydrazide | 123-33-1 | 99.9% | C4H4N2O2 | 50 G
SDP
Catalog No. 5000434751
Encompass_Preferred
Maleic hydrazide is a systemic plant growth regulator and herbicide, known for inhibiting nucleic acid and protein synthesis. It exhibits low acute toxicity via oral, dermal, and inhalation routes, and has shown minimal cytotoxicity on Vero cells compared to Hep2 and HepG2 cells.
  • Systemic plant growth regulator and herbicide
  • Inhibits nucleic acid and protein synthesis
  • Low acute toxicity
  • Minimal cytotoxicity on Vero cells
Catalog No. 50-004-34751 Supplier Medchemexpress LLC Supplier No. HY5935450G
Maleic hydrazide is a systemic plant growth regulator and herbicide, known for inhibiting nucleic acid and protein synthesis. It exhibits low acute toxicity via oral, dermal, and inhalation routes, and has shown minimal cytotoxicity on Vero cells compared to Hep2 and HepG2 cells.
  • Systemic plant growth regulator and herbicide
  • Inhibits nucleic acid and protein synthesis
  • Low acute toxicity
  • Minimal cytotoxicity on Vero cells