Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maleic hydrazide, 97%, Thermo Scientific™

Catalog No. AAA1253136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1253136 500 g
AAA1253122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AAA1253136 Supplier Thermo Scientific Chemicals Supplier No. A1253136

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Maleic hydrazide is a synthetic plant growth regulator that is widely used for controlling growth and to induce dormancy in some crops.

Solubility
Soluble in water (4.5 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-33-1
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.088
MDL Number MFCD00006665
InChI Key BGRDGMRNKXEXQD-UHFFFAOYSA-N
Synonym pyridazine-3,6-diol, maleic hydrazide, 3,6-dihydroxypyridazine, 3,6-pyridazinediol, stuntman, maleic acid hydrazide, vondalhyde, 3,6-pyridazinedione, 1,2-dihydro, 1,2-dihydro-3,6-pyridazinedione, antergon
PubChem CID 21954
ChEBI CHEBI:81771
IUPAC Name 1,2-dihydropyridazine-3,6-dione
SMILES C1=CC(=O)NNC1=O

Specifications

Melting Point ∼305°C (decomposition)
Density 1.6
Flash Point 300°C (572°F)
Quantity 500 g
Beilstein 112223
Merck Index 14,5705
Solubility Information Soluble in water (4.5g/L).
Formula Weight 112.09
Percent Purity 97%
Chemical Name or Material Maleic hydrazide
Hazard Category H315-H319-H335-H341-H351
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 204-619-9
RTECSNumber UR5950000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.