Learn More
Medchemexpress LLC Maleic hydrazide-d2 | 2398483-97-9 | 99.3% | 114.10 g/mol | C4H2D2N2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY59354S10MG
Encompass_Preferred
Maleic hydrazide-d2 is the deuterium-labeled form of maleic hydrazide supplied for research use. It is used as a plant growth regulator and herbicide and is suitable as an isotopically labeled internal standard or tracer for analytical methods such as NMR, GC-MS, and LC-MS.
  • Deuterium-labeled compound (d2) for tracer and standard use.
  • Molecular formula C4H2D2N2O2 and molecular weight 114.10 g/mol.
  • High purity suitable for analytical applications.
  • Offered in small research quantities for laboratory workflows.
  • Store powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).
  • Intended for research use only; not for clinical or patient use.
Catalog No. 50-002-98468