Learn More
Medchemexpress LLC Maleic hydrazide-d2 | 2398483-97-9 | 99.3% | C4H2D2N2O2 | 1 MG
SDP
Catalog No. 5000434754
Encompass_Preferred
Maleic hydrazide-d2 is the deuterium labeled Maleic hydrazide. Maleic hydrazide is widely used as a systemic plant growth regulator and herbicide, and it functions as an inhibitor of nucleic acid and protein synthesis.
  • This compound can be utilized as a tracer.
  • It can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, including deuterium, have been incorporated into drug molecules primarily for quantitation as tracers during the drug development process.
  • Deuteration is notable for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-34754 Supplier Medchemexpress LLC Supplier No. HY59354S1MG
Maleic hydrazide-d2 is the deuterium labeled Maleic hydrazide. Maleic hydrazide is widely used as a systemic plant growth regulator and herbicide, and it functions as an inhibitor of nucleic acid and protein synthesis.
  • This compound can be utilized as a tracer.
  • It can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, including deuterium, have been incorporated into drug molecules primarily for quantitation as tracers during the drug development process.
  • Deuteration is notable for its potential to influence the pharmacokinetic and metabolic profiles of drugs.