Learn More
Medchemexpress LLC Maleic hydrazide-d2 | 2398483-97-9 | 99.3% | C4H2D2N2O2 | 5 MG
SDP
Catalog No. 5000434755
Encompass_Preferred
Maleic hydrazide-d2 is a deuterium-labeled form of Maleic hydrazide, a systemic plant growth regulator and herbicide. It inhibits nucleic acid and protein synthesis and can be used as a tracer or internal standard for quantitative analysis in techniques such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-34755 Supplier Medchemexpress LLC Supplier No. HY59354S5MG
Maleic hydrazide-d2 is a deuterium-labeled form of Maleic hydrazide, a systemic plant growth regulator and herbicide. It inhibits nucleic acid and protein synthesis and can be used as a tracer or internal standard for quantitative analysis in techniques such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may impact pharmacokinetic and metabolic profiles of drugs