Learn More
Medchemexpress LLC Maleimide | 541-59-3 | 97.07 | 1 ML
SDP
Catalog No. 5000460398
Encompass_Preferred
Maleimide is utilized in cancer research for the production of antibody-drug conjugates (ADCs) and for preparing fluorogenic probes specifically designed for detecting thiol analytes. It binds rapidly and covalently to the sulfhydryl group of cysteine residues.
  • Used in drug delivery systems for improved cell uptake efficiency
  • Exhibits antitumor activity in mouse models
  • Has a purity of 99.9%
  • Appears as a white to light yellow solid
  • Features a molecular formula of C4H3NO2
  • Has a molecular weight of 97.07
Catalog No. 50-004-60398 Supplier Medchemexpress LLC Supplier No. HYW00732410MM/1ML
Add to Cart
Add to Cart
Maleimide is utilized in cancer research for the production of antibody-drug conjugates (ADCs) and for preparing fluorogenic probes specifically designed for detecting thiol analytes. It binds rapidly and covalently to the sulfhydryl group of cysteine residues.
  • Used in drug delivery systems for improved cell uptake efficiency
  • Exhibits antitumor activity in mouse models
  • Has a purity of 99.9%
  • Appears as a white to light yellow solid
  • Features a molecular formula of C4H3NO2
  • Has a molecular weight of 97.07